Assay Details
| Gene Symbol: ZNF41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324151.1 | NP_001311080.1 | 2 - 3 | 525 | 138 | |
| NM_001324153.1 | NP_001311082.1 | 2 - 3 | 435 | 138 | ||
| XM_006724555.3 | XP_006724618.1 | 2 - 3 | 819 | 138 | ||
| XM_017029816.1 | XP_016885305.1 | 2 - 3 | 358 | 138 | ||
| GenBank mRNA | AJ010018.1 | - | 1 - 2 | 138 | 138 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ010018.1, NM_001324151.1, NM_001324153.1, XM_006724555.3, XM_017029816.1 transcripts.
Set Membership: |
|

ZNF41
Genomic Map