| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013208.hg.1 |
TC0X000996.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013208.hg.1 |
ENST00000313116.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013208.hg.1 |
ENST00000377065.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013208.hg.1 |
ENST00000397050.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013208.hg.1 |
ENST00000465311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013208.hg.1 |
NM_007130.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013208.hg.1 |
NM_153380.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013212.hg.1 |
TC0X000996.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013212.hg.1 |
ENST00000313116.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013212.hg.1 |
ENST00000377065.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013212.hg.1 |
ENST00000397050.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013212.hg.1 |
ENST00000465311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013212.hg.1 |
NM_007130.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013212.hg.1 |
NM_153380.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
ENST00000313116.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
ENST00000377065.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
ENST00000432977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
ENST00000465311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
NM_007130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
NM_153380.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
ENST00000313116.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
ENST00000377065.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
ENST00000432977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
ENST00000465311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
NM_007130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
NM_153380.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165215.hg.1 |
TC0X00009569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
ENST00000313116.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
ENST00000377065.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
ENST00000432977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
ENST00000465311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
NM_007130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
NM_153380.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
ENST00000313116.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
ENST00000377065.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
ENST00000432977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
ENST00000465311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
NM_007130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
NM_153380.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165218.hg.1 |
TC0X00009569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
ENST00000313116.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
ENST00000377065.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
ENST00000432977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
ENST00000465311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
NM_007130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
NM_153380.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
ENST00000313116.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
ENST00000377065.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
ENST00000432977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
ENST00000465311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
NM_007130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
NM_153380.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165219.hg.1 |
TC0X00009569.hg.1 |
Clariom D TC |