Assay Details
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014878.4 | NP_055693.4 | 1 - 2 | 84 | 152 | |
GenBank mRNA | AK225953.1 | - | 1 - 2 | 49 | 152 | |
AK315531.1 | - | 1 - 2 | 63 | 152 | ||
AL832239.1 | - | 1 - 2 | 50 | 152 | ||
AL832245.1 | - | 1 - 2 | 47 | 152 | ||
DA358022.1 | - | 1 - 2 | 84 | 152 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000874.hg
Additional Information:
This assay is in the 5'UTR region of the AK315531.1,NM_014878.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09011638.hg.1 | TC09000874.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09011638.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09011638.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09011636.hg.1 | TC09000874.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09011636.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09011636.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900164605.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164605.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900164605.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164605.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900164605.hg.1 | TC0900009404.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164607.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164607.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900164607.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164607.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900164607.hg.1 | TC0900009404.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 175386 | - | - |
