Assay Details
Gene Symbol: LRRC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098519.1 | NP_001091989.1 | 1 - 2 | 171 | 71 | |
GenBank mRNA | DA951767.1 | - | 1 - 2 | 171 | 71 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000970.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12012686.hg.1 | ENST00000289014.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012686.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012686.hg.1 | ENST00000425921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012686.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12012686.hg.1 | TC12000970.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200168193.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200168193.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200168193.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200168193.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200168193.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200168193.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200168193.hg.1 | TC1200009198.hg.1 | Clariom D TC |
