Assay Details
Gene Symbol: LRRC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098519.1 | NP_001091989.1 | 3 - 4 | 549 | 87 | |
NM_152759.4 | NP_689972.3 | 3 - 4 | 438 | 87 | ||
XM_017019124.1 | XP_016874613.1 | 3 - 4 | 557 | 87 | ||
XM_017019125.1 | XP_016874614.1 | 3 - 4 | 557 | 87 | ||
GenBank mRNA | BC029245.2 | - | 2 - 3 | 373 | 5169080 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000970.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC029245.2,NM_152759.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12012691.hg.1 | ENST00000289014.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012691.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012691.hg.1 | ENST00000425921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012691.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12012691.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12012691.hg.1 | TC12000970.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200168200.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200168200.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200168200.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200168200.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200168200.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200168200.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200168200.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200168200.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200168200.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200168200.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200168200.hg.1 | TC1200009198.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
