Assay Details
| Gene Symbol: NFE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261461.1 | NP_001248390.1 | 2 - 3 | 364 | 84 | |
| GenBank mRNA | AK313207.1 | - | 2 - 3 | 342 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK313207.1, NM_001261461.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nucleosome disassembly
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
multicellular organism development
blood coagulation
hemostasis
blood circulation
positive regulation of transcription, DNA-templated
transcription regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
sequence-specific DNA binding
protein N-terminus binding
WW domain binding
nucleosome disassembly
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
multicellular organism development
blood coagulation
hemostasis
blood circulation
positive regulation of transcription, DNA-templated
transcription regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
sequence-specific DNA binding
protein N-terminus binding
WW domain binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183164.hg.1 | TC1200010835.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010835.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | ENST00000312156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | ENST00000435572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | ENST00000540264.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | ENST00000553070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | ENST00000553198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | NM_001136023.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | NM_001261461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | NM_006163.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183163.hg.1 | TC1200010835.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 205836 | - | - |

NFE2
Genomic Map