Assay Details
| Gene Symbol: LOC643659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK056971.1 | - | 1 | 397 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | crooked neck pre-mRNA splicing factor 1 | CRNKL1 |
CLF, CRN, Clf1, HCRN, MSTP021, SYF3 |
-19.3kb | Sense | Chr. 20 - 20034361 - 20056046 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929575 | LOC101929575 | 178.4kb | Sense | Chr. 20 - 20254830 - 20267834 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC643659
Genomic Map