Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006337.hg.1 |
ENST00000255448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006337.hg.1 |
ENST00000360631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006337.hg.1 |
ENST00000379893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006337.hg.1 |
ENST00000399319.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006337.hg.1 |
ENST00000539451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006337.hg.1 |
NM_001195415.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006337.hg.1 |
NM_001195416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006337.hg.1 |
NM_004734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006337.hg.1 |
TC13000563.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
ENST00000255448.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
ENST00000360631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
ENST00000379893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
ENST00000615680.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
NM_001195415.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
NM_001195416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
NM_004734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
ENST00000255448.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
ENST00000360631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
ENST00000379893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
ENST00000615680.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
NM_001195415.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
NM_001195416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
NM_004734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158023.hg.1 |
TC1300008609.hg.1 |
Clariom D TC |