Assay Details
Gene Symbol: SPRYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127482.2 | NP_001120954.1 | 1 - 2 | 370 | 89 | |
GenBank mRNA | AK290576.1 | - | 1 - 2 | 209 | 89 | |
BC022519.1 | - | 1 - 2 | 176 | 4792366 | 89 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000667.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13007695.hg.1 | ENST00000361840.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007695.hg.1 | ENST00000378195.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007695.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007695.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007695.hg.1 | TC13000667.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13007699.hg.1 | ENST00000361840.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007699.hg.1 | ENST00000378195.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007699.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007699.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007699.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007699.hg.1 | TC13000667.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13007700.hg.1 | ENST00000378195.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007700.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007700.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007700.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007700.hg.1 | TC13000667.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000492258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000492258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007698.hg.1 | ENST00000361840.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007698.hg.1 | ENST00000378195.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007698.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007698.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007698.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007698.hg.1 | TC13000667.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1300160304.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160304.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160304.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160304.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160304.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160304.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160304.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160304.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160304.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160304.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160304.hg.1 | TC1300008936.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300160310.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160310.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160310.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160310.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160310.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160310.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160310.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160310.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160310.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160310.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160310.hg.1 | TC1300008936.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300160311.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160311.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160311.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160311.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160311.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160311.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160311.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160311.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160311.hg.1 | TC1300008936.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160308.hg.1 | TC1300008936.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160307.hg.1 | TC1300008936.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000361840.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000378195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000492258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000492258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | ENST00000613924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_001127482.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NM_020456.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NR_023351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008936.hg.1 | NR_023351.2 | Refseq |
