Assay Details
Gene Symbol: C18orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011525822.1 | XP_011524124.1 | 1 - 2 | 161 | 113 | |
XM_017025555.1 | XP_016881044.1 | 1 - 2 | 157 | 113 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011525822.1, XM_017025555.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010235.hg.1 | ENST00000392363.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010235.hg.1 | ENST00000392363.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
