Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005220.hg.1 |
ENST00000224721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005220.hg.1 |
ENST00000398855.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005220.hg.1 |
ENST00000398860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005220.hg.1 |
NM_001171930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005220.hg.1 |
NM_022124.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005220.hg.1 |
TC10000442.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
ENST00000224721.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
ENST00000398809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
ENST00000616684.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
ENST00000622827.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
NM_001171930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
NM_022124.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
ENST00000224721.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
ENST00000398809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
ENST00000616684.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
ENST00000622827.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
NM_001171930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
NM_022124.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154047.hg.1 |
TC1000007959.hg.1 |
Clariom D TC |