Assay Details
| Gene Symbol: DACH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011534939.2 | XP_011533241.1 | 4 - 5 | 2585 | 85 | |
| XM_011534940.2 | XP_011533242.1 | 4 - 5 | 2585 | 85 | ||
| XM_017020396.1 | XP_016875885.1 | 3 - 4 | 579 | 85 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
suckling behavior
multicellular organism development
respiratory gaseous exchange
cell proliferation
negative regulation of transcription by competitive promoter binding
negative regulation of cell migration
regulation of nuclear cell cycle DNA replication
negative regulation of transcription, DNA-templated
development of primary female sexual characteristics
negative regulation of fibroblast proliferation
RNA polymerase II transcriptional preinitiation complex assembly
negative regulation of cell proliferation involved in contact inhibition
negative regulation of DNA biosynthetic process
transcription factor activity, RNA polymerase II core promoter sequence-specific binding involved in preinitiation complex assembly
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
protein binding
negative regulation of transcription from RNA polymerase II promoter
suckling behavior
multicellular organism development
respiratory gaseous exchange
cell proliferation
negative regulation of transcription by competitive promoter binding
negative regulation of cell migration
regulation of nuclear cell cycle DNA replication
negative regulation of transcription, DNA-templated
development of primary female sexual characteristics
negative regulation of fibroblast proliferation
RNA polymerase II transcriptional preinitiation complex assembly
negative regulation of cell proliferation involved in contact inhibition
negative regulation of DNA biosynthetic process
transcription factor activity, RNA polymerase II core promoter sequence-specific binding involved in preinitiation complex assembly
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000611519.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000613252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000619232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | ENST00000620444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_004392.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080759.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009218.hg.1 | NM_080760.5 | Refseq |

DACH1
Genomic Map