Assay Details
Gene Symbol: ENTPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249.3 | NP_001240.1 | 3 - 4 | 242 | 86 | |
NM_001321988.1 | NP_001308917.1 | 3 - 4 | 1018 | 86 | ||
XM_005268224.3 | XP_005268281.1 | 3 - 4 | 204 | 86 | ||
XM_006720325.3 | XP_006720388.1 | 3 - 4 | 245 | 86 | ||
XM_006720326.2 | XP_006720389.1 | 3 - 4 | 245 | 86 | ||
XM_017021817.1 | XP_016877306.1 | 3 - 4 | 245 | 86 | ||
GenBank mRNA | AF039918.1 | - | 2 - 3 | 177 | 86 | |
AY430094.1 | - | 2 - 3 | 183 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF039918.1, AY430094.1, NM_001249.3, NM_001321988.1, XM_005268224.3, XM_006720325.3, XM_006720326.2, XM_017021817.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
