Assay Details
| Gene Symbol: LAMTOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243736.1 | NP_001230665.1 | 2 - 3 | 260 | 89 | |
| NM_021970.3 | NP_068805.1 | 2 - 3 | 260 | 89 | ||
| GenBank mRNA | AF130115.1 | - | 1 - 2 | 60 | 89 | |
| AF201947.1 | - | 2 - 3 | 170 | 89 | ||
| AK223311.1 | - | 2 - 3 | 151 | 89 | ||
| AK311755.1 | - | 2 - 3 | 121 | 89 | ||
| AL544981.3 | - | 2 - 3 | 90 | 89 | ||
| BC026245.1 | - | 2 - 3 | 153 | 4808855 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF130115.1, AF201947.1, AK311755.1, BC026245.1, NM_001243736.1, NM_021970.3 transcripts.
Set Membership: |
|

LAMTOR3
Genomic Map