Assay Details
| Gene Symbol: LAMTOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243736.1 | NP_001230665.1 | 3 - 4 | 293 | 73 | |
| NM_021970.3 | NP_068805.1 | 3 - 4 | 293 | 73 | ||
| GenBank mRNA | AF130115.1 | - | 2 - 3 | 93 | 73 | |
| AF201947.1 | - | 3 - 4 | 203 | 73 | ||
| AK223311.1 | - | 3 - 4 | 184 | 73 | ||
| AK311755.1 | - | 3 - 4 | 154 | 73 | ||
| AL544981.3 | - | 3 - 4 | 123 | 73 | ||
| BC026245.1 | - | 3 - 4 | 186 | 4808855 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LAMTOR3
Genomic Map