Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025809.hg.1 |
ENST00000327551.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025809.hg.1 |
ENST00000419234.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025809.hg.1 |
ENST00000539060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025809.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025809.hg.1 |
TC12001974.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025810.hg.1 |
ENST00000327551.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025810.hg.1 |
ENST00000419234.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025810.hg.1 |
ENST00000539060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025810.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025810.hg.1 |
TC12001974.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200192834.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192834.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192834.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192834.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192834.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192834.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192834.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192834.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192834.hg.1 |
TC1200011946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200192836.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192836.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192836.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192836.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192836.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192836.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192836.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192836.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192836.hg.1 |
TC1200011946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000327551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000419234.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
ENST00000547043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011946.hg.1 |
NM_006768.4 |
Refseq |