Assay Details
| Gene Symbol: PLD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308174.1 | NP_001295103.1 | 4 - 5 | 682 | 107 | |
| NM_138790.3 | NP_620145.2 | 4 - 5 | 644 | 107 | ||
| XM_011536411.2 | XP_011534713.1 | 4 - 5 | 692 | 107 | ||
| XM_017020965.1 | XP_016876454.1 | 4 - 5 | 581 | 107 | ||
| GenBank mRNA | AK295352.1 | - | 4 - 5 | 637 | 107 | |
| AK296188.1 | - | 4 - 5 | 682 | 107 | ||
| AY358843.1 | - | 4 - 5 | 610 | 107 | ||
| BC015003.1 | - | 4 - 5 | 573 | 4853026 | 107 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000828.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14009051.hg.1 | ENST00000392593.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009051.hg.1 | ENST00000540372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009051.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009051.hg.1 | TC14000828.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000472702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000472702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000472702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000472702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000472702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000472901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000472901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000472901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000472901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000472901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000553861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000553861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000553861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000553861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000553861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400160629.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400160629.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400160629.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400160629.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400160629.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400160629.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400160629.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400160629.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400160629.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400160629.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400160629.hg.1 | TC1400008460.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000392593.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000472702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000472702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000472702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000472702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000472702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000472901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000472901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000472901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000472901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000472901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000540372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000553861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000553861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000553861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000553861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000553861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | ENST00000557573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_001308174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008460.hg.1 | NM_138790.3 | Refseq |

PLD4
Genomic Map