Assay Details
| Gene Symbol: FCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK314867.1 | - | 4 - 5 | 346 | 136 | |
| BC020635.1 | - | 4 - 5 | 328 | 4717731 | 136 | |
| CR749452.1 | - | 4 - 5 | 314 | 136 | ||
| D83920.1 | - | 4 - 5 | 302 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011918.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181494.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181494.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181494.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181494.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181494.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181494.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181494.hg.1 | TC0900011918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900181495.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181495.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181495.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181495.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181495.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181495.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181495.hg.1 | TC0900011918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900181496.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181496.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181496.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181496.hg.1 | ENST00000371806.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181496.hg.1 | ENST00000616356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181496.hg.1 | NM_002003.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181496.hg.1 | TC0900011918.hg.1 | Clariom D TC |

FCN1
Genomic Map