| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006931.hg.1 |
TC08000506.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006931.hg.1 |
ENST00000263849.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006931.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000521176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000521176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000521176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000521176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000521873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000521873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000521873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
ENST00000521873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155315.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155315.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155315.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155315.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155315.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155315.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155315.hg.1 |
TC0800008032.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000263849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000519307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000521176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000521176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000521176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000521176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000521873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000521873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000521873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
ENST00000521873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008032.hg.1 |
NM_016010.2 |
Refseq |