Assay Details
| Gene Symbol: TBXA2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001060.5 | NP_001051.1 | 1 - 2 | 303 | 52 | |
| NM_201636.2 | NP_963998.2 | 1 - 2 | 303 | 52 | ||
| GenBank mRNA | BC028027.1 | - | 1 - 2 | 303 | 52 | |
| U11271.1 | - | 1 - 2 | 128 | 52 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001046.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC028027.1,NM_001060.5,NM_201636.2,U11271.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
inflammatory response
G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
response to nutrient
second-messenger-mediated signaling
response to lipopolysaccharide
thromboxane A2 signaling pathway
response to drug
positive regulation of GTPase activity
positive regulation of angiogenesis
positive regulation of blood pressure
positive regulation of vasoconstriction
positive regulation of smooth muscle contraction
thromboxane A2 receptor activity
guanyl-nucleotide exchange factor activity
protein binding
inflammatory response
G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
response to nutrient
second-messenger-mediated signaling
response to lipopolysaccharide
thromboxane A2 signaling pathway
response to drug
positive regulation of GTPase activity
positive regulation of angiogenesis
positive regulation of blood pressure
positive regulation of vasoconstriction
positive regulation of smooth muscle contraction
thromboxane A2 receptor activity
guanyl-nucleotide exchange factor activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19014253.hg.1 | ENST00000375190.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014253.hg.1 | ENST00000411851.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014253.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014253.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014253.hg.1 | TC19001046.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014250.hg.1 | ENST00000375190.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014250.hg.1 | ENST00000411851.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014250.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014250.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014250.hg.1 | TC19001046.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014252.hg.1 | ENST00000375190.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014252.hg.1 | ENST00000411851.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014252.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014252.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014252.hg.1 | TC19001046.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000587717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000587717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174097.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174097.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174097.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174097.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174097.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174097.hg.1 | TC1900009297.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174097.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174097.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174097.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174097.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174097.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174098.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174098.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174098.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174098.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174098.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174098.hg.1 | TC1900009297.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174098.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174098.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174098.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174098.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174098.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174101.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174101.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174101.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174101.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174101.hg.1 | TC1900009297.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174101.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174101.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174101.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174101.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000587717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000587717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | ENST00000589966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009297.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174102.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174102.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174102.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174102.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174102.hg.1 | TC1900009297.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174102.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174102.hg.1 | ENST00000411851.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174102.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174102.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174103.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174103.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174103.hg.1 | NM_201636.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174103.hg.1 | TC1900009297.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174103.hg.1 | ENST00000375190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174103.hg.1 | NM_001060.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174103.hg.1 | NM_201636.2 | Refseq |

TBXA2R
Genomic Map