Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013114.hg.1 |
TC01000823.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013114.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013114.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013114.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000496322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000496322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
ENST00000496322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163677.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163677.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163677.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163677.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163677.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163677.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163677.hg.1 |
TC0100008924.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000370563.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000496322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000496322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
ENST00000496322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NM_012128.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008924.hg.1 |
NR_024602.1 |
Refseq |