Assay Details
Gene Symbol: CLCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012128.3 | NP_036260.2 | 1 - 2 | 203 | 154 | |
NR_024602.1 | 1 - 2 | 203 | 154 | |||
GenBank mRNA | AF127035.1 | - | 1 - 2 | 187 | 154 | |
AK000072.1 | - | 1 - 2 | 201 | 154 | ||
AK307785.1 | - | 1 - 2 | 147 | 154 | ||
AY358470.1 | - | 1 - 2 | 183 | 154 | ||
BC113687.1 | - | 1 - 2 | 198 | 8322739 | 154 | |
BC113689.1 | - | 1 - 2 | 197 | 8322741 | 154 | |
DA761219.1 | - | 1 - 2 | 265 | 154 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000823.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01013100.hg.1 | TC01000823.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01013100.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01013100.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01013100.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01013102.hg.1 | TC01000823.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01013102.hg.1 | ENST00000263723.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01013102.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01013102.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01013102.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01013101.hg.1 | TC01000823.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01013101.hg.1 | ENST00000263723.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01013101.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01013101.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01013101.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000496322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000496322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | ENST00000496322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000496322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000496322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | ENST00000496322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008924.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163665.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100163665.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163665.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163665.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100163665.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163665.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163665.hg.1 | TC0100008924.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100163666.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100163666.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163666.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163666.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100163666.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163666.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163666.hg.1 | TC0100008924.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100163664.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100163664.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163664.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163664.hg.1 | ENST00000370563.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100163664.hg.1 | NM_012128.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163664.hg.1 | NR_024602.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163664.hg.1 | TC0100008924.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 213600 | - | - |
