Assay Details
| Gene Symbol: CLCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF039400.1 | - | 1 - 2 | 90 | 151 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF039400.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |

CLCA1
Genomic Map