Assay Details
| Gene Symbol: CLCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285.3 | NP_001276.2 | 2 - 3 | 438 | 77 | |
| GenBank mRNA | AF039400.1 | - | 3 - 4 | 660 | 77 | |
| AF127036.1 | - | 2 - 3 | 313 | 77 | ||
| AK291999.1 | - | 2 - 3 | 354 | 77 | ||
| AK314375.1 | - | 2 - 3 | 356 | 77 | ||
| DA428844.1 | - | 2 - 3 | 438 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000822.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013077.hg.1 | TC01000822.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013077.hg.1 | ENST00000234701.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013077.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013077.hg.1 | ENST00000495894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013077.hg.1 | ENST00000539889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013077.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013075.hg.1 | TC01000822.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013075.hg.1 | ENST00000234701.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013075.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013075.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100163639.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100163639.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100163639.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100163639.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100163639.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100163639.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100163639.hg.1 | TC0100008922.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000234701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | ENST00000394711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008922.hg.1 | NM_001285.3 | Refseq |

CLCA1
Genomic Map