Assay Details
Gene Symbol: KCNH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144633.2 | NP_653234.2 | 12 - 13 | 2276 | 95 | |
XM_011533347.2 | XP_011531649.1 | 8 - 9 | 1404 | 95 | ||
XM_017005698.1 | XP_016861187.1 | 12 - 13 | 2347 | 95 | ||
XM_017005699.1 | XP_016861188.1 | 11 - 12 | 2013 | 95 | ||
XM_017005700.1 | XP_016861189.1 | 10 - 11 | 1981 | 95 | ||
XM_017005701.1 | XP_016861190.1 | 13 - 14 | 2417 | 95 | ||
XM_017005702.1 | XP_016861191.1 | 10 - 11 | 2862 | 95 | ||
GenBank mRNA | AB209053.1 | - | 12 - 13 | 3199 | 95 | |
AK294417.1 | - | 3 - 4 | 681 | 95 | ||
AY053503.1 | - | 12 - 13 | 2276 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000110.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
