Assay Details
| Gene Symbol: SEC22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012430.4 | NP_036562.2 | 5 - 6 | 731 | 150 | |
| XM_011512673.2 | XP_011510975.1 | 5 - 6 | 2247 | 150 | ||
| XM_011512675.2 | XP_011510977.1 | 6 - 7 | 898 | 150 | ||
| XM_011512676.2 | XP_011510978.1 | 5 - 6 | 787 | 150 | ||
| GenBank mRNA | AF100749.1 | - | 5 - 6 | 713 | 150 | |
| AK057587.1 | - | 4 - 5 | 1547 | 150 | ||
| AK315771.1 | - | 5 - 6 | 700 | 150 | ||
| BC007122.1 | - | 4 - 5 | 679 | 3459649 | 150 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003364.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03011546.hg.1 | TC03003364.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011546.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011546.hg.1 | ENST00000481965.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011546.hg.1 | ENST00000492595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011546.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300205193.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300205193.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300205193.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300205193.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300205193.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300205193.hg.1 | ENST00000309934.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300205193.hg.1 | ENST00000473494.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300205193.hg.1 | ENST00000481965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300205193.hg.1 | ENST00000492595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300205193.hg.1 | NM_012430.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300205193.hg.1 | TC0300013865.hg.1 | Clariom D TC |

SEC22A
Genomic Map