Assay Details
Gene Symbol: ABI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135186.1 | NP_001128658.1 | 6 - 7 | 1284 | 99 | |
NM_016428.2 | NP_057512.1 | 6 - 7 | 1302 | 99 | ||
XM_017024721.1 | XP_016880210.1 | 6 - 7 | 944 | 99 | ||
GenBank mRNA | AB037886.1 | - | 6 - 7 | 1302 | 99 | |
AL136709.1 | - | 6 - 7 | 1103 | 99 | ||
BC007780.2 | - | 6 - 7 | 808 | 4309524 | 99 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000640.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007994.hg.1 | ENST00000225941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007994.hg.1 | ENST00000419580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007994.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007994.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007994.hg.1 | TC17000640.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160625.hg.1 | TC1700008194.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000225941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000419580.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000571035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | ENST00000573347.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_001135186.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008194.hg.1 | NM_016428.2 | Refseq |
