Assay Details
| Gene Symbol: UBE2Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023079.4 | NP_075567.2 | 5 - 6 | 943 | 100 | |
| GenBank mRNA | AB025426.1 | - | 4 - 5 | 484 | 100 | |
| AK023917.1 | - | 4 - 5 | 481 | 100 | ||
| AK024030.1 | - | 6 - 7 | 798 | 100 | ||
| AK091644.1 | - | 3 - 4 | 2074 | 100 | ||
| AK122770.1 | - | 5 - 6 | 841 | 100 | ||
| AK130303.1 | - | 6 - 7 | 801 | 100 | ||
| AK296865.1 | - | 6 - 7 | 834 | 100 | ||
| AL713782.1 | - | 1 - 2 | 99 | 100 | ||
| BC015169.1 | - | 4 - 5 | 470 | 100 | ||
| BC015890.1 | - | 5 - 6 | 811 | 3915698 | 100 | |
| CR457322.1 | - | 4 - 5 | 484 | 100 | ||
| EF623992.1 | - | 5 - 6 | 808 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000634.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17007927.hg.1 | ENST00000360943.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007927.hg.1 | ENST00000405215.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007927.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007927.hg.1 | TC17000634.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000514948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000514948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | ENST00000514948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000508468.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000511373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000514948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000514948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | ENST00000514948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008180.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000514948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000360943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000504684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000506271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000506498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000513342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | ENST00000514948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | NM_023079.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160528.hg.1 | TC1700008180.hg.1 | Clariom D TC |

UBE2Z
Genomic Map