Assay Details
| Gene Symbol: IL18RAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003853.3 | NP_003844.1 | 3 - 4 | 559 | 130 | |
| GenBank mRNA | AF077346.1 | - | 3 - 4 | 554 | 130 | |
| AK300026.1 | - | 3 - 4 | 659 | 130 | ||
| BC069630.1 | - | 1 - 2 | 156 | 7262472 | 130 | |
| BC106765.2 | - | 2 - 3 | 283 | 40026249 | 130 | |
| BC137474.1 | - | 1 - 2 | 99 | 9021474 | 130 | |
| BC137475.1 | - | 1 - 2 | 99 | 9021475 | 130 | |
| DQ116957.1 | - | 1 - 2 | 71 | 130 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000623.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02009943.hg.1 | TC02000623.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009943.hg.1 | ENST00000264260.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009943.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009943.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009944.hg.1 | TC02000623.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009944.hg.1 | ENST00000264260.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009944.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009945.hg.1 | TC02000623.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009945.hg.1 | ENST00000264260.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009945.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009942.hg.1 | TC02000623.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009942.hg.1 | ENST00000264260.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009942.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009942.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160646.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160646.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160646.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160646.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160646.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160646.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160646.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160646.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160646.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160646.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160646.hg.1 | TC0200008675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200160645.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160645.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160645.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160645.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160645.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160645.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160645.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160645.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160645.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160645.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160645.hg.1 | TC0200008675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008675.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160648.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160648.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160648.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160648.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160648.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160648.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160648.hg.1 | TC0200008675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200160644.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160644.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160644.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160644.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160644.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160644.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160644.hg.1 | ENST00000409369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160644.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160644.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160644.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160644.hg.1 | TC0200008675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200160647.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160647.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160647.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160647.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160647.hg.1 | ENST00000264260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160647.hg.1 | ENST00000450855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160647.hg.1 | ENST00000497795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160647.hg.1 | NM_003853.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160647.hg.1 | TC0200008675.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 104771 | - | - |

IL18RAP
Genomic Map