Assay Details
| Gene Symbol: PARP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005264779.4 | XP_005264836.2 | 2 - 3 | 542 | 93 | |
| GenBank mRNA | BG818516.1 | - | 2 - 3 | 479 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005264779.4 transcripts.
Set Membership: |
|

PARP3
Genomic Map