Assay Details
| Gene Symbol: POLR3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018050.3 | NP_001018060.1 | 3 - 4 | 678 | 129 | |
| NM_001282884.1 | NP_001269813.1 | 4 - 5 | 406 | 129 | ||
| NM_001282885.1 | NP_001269814.1 | 4 - 5 | 365 | 129 | ||
| NM_138338.4 | NP_612211.1 | 4 - 5 | 372 | 129 | ||
| GenBank mRNA | AB051452.1 | - | 3 - 4 | 533 | 129 | |
| AK026524.1 | - | 4 - 5 | 352 | 129 | ||
| AY092087.1 | - | 4 - 5 | 350 | 129 | ||
| BC088367.1 | - | 3 - 4 | 541 | 30385483 | 129 | |
| BU173504.1 | - | 3 - 4 | 320 | 129 | ||
| CR456459.1 | - | 4 - 5 | 406 | 129 | ||
| DA449644.1 | - | 4 - 5 | 387 | 129 | ||
| DA504810.1 | - | 4 - 5 | 354 | 129 | ||
More Information
Set Membership: |
|

POLR3H
Genomic Map