Assay Details
| Gene Symbol: TSC22D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130136.1 | 2 - 3 | 3044 | 89 | ||
| GenBank mRNA | AF201292.1 | - | 2 - 3 | 200 | 89 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF201292.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000492828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000492828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300205614.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300205614.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300205614.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300205614.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300205614.hg.1 | TC0300013888.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000492828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000492828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |

TSC22D2
Genomic Map