Assay Details
| Gene Symbol: PITPNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284278.1 | NP_001271207.1 | 10 - 11 | 862 | 67 | |
| NM_012399.4 | NP_036531.1 | 10 - 11 | 884 | 67 | ||
| GenBank mRNA | AK302367.1 | - | 10 - 11 | 848 | 67 | |
| AK313064.1 | - | 10 - 11 | 834 | 67 | ||
| BC018704.1 | - | 10 - 11 | 808 | 4643509 | 67 | |
| CR456541.1 | - | 10 - 11 | 836 | 67 | ||
| D30037.1 | - | 10 - 11 | 815 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000621.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000460566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000460566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000460566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000460566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000465179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000465179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000465179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000465179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000471825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000471825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000471825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000493007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000493007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000493007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000493007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |

PITPNB
Genomic Map