Assay Details
| Gene Symbol: PITPNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284278.1 | NP_001271207.1 | 1 - 2 | 117 | 64 | |
| XM_011530052.2 | XP_011528354.1 | 1 - 2 | 152 | 64 | ||
| XM_017028707.1 | XP_016884196.1 | 1 - 2 | 112 | 64 | ||
| GenBank mRNA | AK302367.1 | - | 1 - 2 | 103 | 64 | |
| DC313909.1 | - | 1 - 2 | 117 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK302367.1, NM_001284278.1, XM_011530052.2, XM_017028707.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000320996.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000335272.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000415296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000436663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000455418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000460566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000460566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000460566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000460566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000465179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000465179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000465179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000465179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000471825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000471825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000471825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000477861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000493007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000493007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000493007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000493007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | ENST00000634017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_001284278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008350.hg.1 | NM_012399.4 | Refseq |

PITPNB
Genomic Map