Assay Details
| Gene Symbol: NMNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297778.1 | NP_001284707.1 | 77 | |||
| NM_001297779.1 | NP_001284708.1 | 77 | ||||
| NM_022787.3 | NP_073624.2 | 77 | ||||
| XM_011541971.2 | XP_011540273.1 | 77 | ||||
| XM_017002107.1 | XP_016857596.1 | 77 | ||||
| XM_017002108.1 | XP_016857597.1 | 77 | ||||
| GenBank mRNA | AF312734.1 | - | 77 | |||
| AF314163.1 | - | 77 | ||||
| AF459819.1 | - | 77 | ||||
| AK026065.1 | - | 77 | ||||
| AK222920.1 | - | 77 | ||||
| AK315640.1 | - | 77 | ||||
| BC014943.1 | - | 4874147 | 77 | |||
| BP309588.1 | - | 77 | ||||
| BP339298.1 | - | 77 | ||||
| BU948893.1 | - | 77 | ||||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000122.hg.1
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
NAD biosynthetic process
response to wounding
NAD metabolic process
'de novo' NAD biosynthetic process from aspartate
ATP generation from poly-ADP-D-ribose
nicotinamide-nucleotide adenylyltransferase activity
nicotinate-nucleotide adenylyltransferase activity
protein binding
ATP binding
NAD biosynthetic process
response to wounding
NAD metabolic process
'de novo' NAD biosynthetic process from aspartate
ATP generation from poly-ADP-D-ribose
nicotinamide-nucleotide adenylyltransferase activity
nicotinate-nucleotide adenylyltransferase activity
protein binding
ATP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01001870.hg.1 | TC01000122.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001870.hg.1 | ENST00000377205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001870.hg.1 | ENST00000403197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001870.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000413148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000413148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000445884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000445884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000492735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000492735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000492735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000492735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000496751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | ENST00000496751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | ENST00000496751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | ENST00000496751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147041.hg.1 | TC0100006801.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100147042.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147042.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147042.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147042.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147042.hg.1 | TC0100006801.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000377205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000413148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000413148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000445884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000445884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000462686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000492735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000492735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000492735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000492735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000496751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | ENST00000496751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006801.hg.1 | NM_022787.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147043.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147043.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147043.hg.1 | ENST00000403197.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147043.hg.1 | NM_001297779.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147043.hg.1 | TC0100006801.hg.1 | Clariom D TC |

NMNAT1
Genomic Map