Assay Details
| Gene Symbol: EXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286441.1 | NP_001273370.1 | 7 - 8 | 866 | 157 | |
| NM_152596.3 | NP_689809.2 | 5 - 6 | 611 | 157 | ||
| XM_006720415.3 | XP_006720478.1 | 6 - 7 | 783 | 157 | ||
| XM_011521298.2 | XP_011519600.1 | 8 - 9 | 1096 | 157 | ||
| XM_011521299.2 | XP_011519601.1 | 8 - 9 | 827 | 157 | ||
| XM_011521300.2 | XP_011519602.1 | 5 - 6 | 355 | 157 | ||
| XM_011521301.2 | XP_011519603.1 | 7 - 8 | 652 | 157 | ||
| XM_017021970.1 | XP_016877459.1 | 5 - 6 | 355 | 157 | ||
| XM_017021971.1 | XP_016877460.1 | 5 - 6 | 353 | 157 | ||
| GenBank mRNA | AK292524.1 | - | 5 - 6 | 517 | 157 | |
| AK302864.1 | - | 7 - 8 | 800 | 157 | ||
| BC030628.2 | - | 5 - 6 | 551 | 4827307 | 157 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001247.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011315.hg.1 | ENST00000314992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011315.hg.1 | ENST00000458580.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011315.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011315.hg.1 | TC15001247.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500166234.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166234.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166234.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166234.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166234.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166234.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166234.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166234.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166234.hg.1 | TC1500009143.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |

EXD1
Genomic Map