Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000517885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000518079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000518079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000518079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000518079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000518079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000522677.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000523304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000523304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000523304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000523304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000523304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000523304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000523304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000523304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000523304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
ENST00000524176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001080416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001144755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010685.hg.1 |
NM_001294282.1 |
Refseq |