Assay Details
Gene Symbol: MYBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080416.3 | NP_001073885.1 | 9 - 10 | 1516 | 97 | |
NM_001144755.2 | NP_001138227.1 | 9 - 10 | 1516 | 97 | ||
NM_001294282.1 | NP_001281211.1 | 9 - 10 | 1513 | 97 | ||
XM_017013456.1 | XP_016868945.1 | 9 - 10 | 1364 | 97 | ||
XM_017013458.1 | XP_016868947.1 | 9 - 10 | 1362 | 97 | ||
GenBank mRNA | AK308490.1 | - | 2 - 3 | 283 | 97 | |
BC101186.1 | - | 9 - 10 | 1171 | 40018967 | 97 | |
BC101188.1 | - | 9 - 10 | 1174 | 40018969 | 97 | |
X13269.1 | - | 9 - 10 | 1209 | 97 | ||
X13294.1 | - | 9 - 10 | 1209 | 97 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001286.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
