Assay Details
| Gene Symbol: DCAF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028482.1 | 7 - 8 | 1056 | 76 | ||
| XM_017004999.1 | XP_016860488.1 | 7 - 8 | 1051 | 76 | ||
| XM_017005000.1 | XP_016860489.1 | 7 - 8 | 1051 | 76 | ||
| XM_017005001.1 | XP_016860490.1 | 7 - 8 | 1051 | 76 | ||
| GenBank mRNA | AK023158.1 | - | 7 - 8 | 687 | 76 | |
| BC120957.1 | - | 2 - 3 | 196 | 76 | ||
| BC144653.1 | - | 7 - 8 | 836 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK023158.1, BC120957.1 transcripts.
Set Membership: |
|

DCAF17
Genomic Map