Assay Details
| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004175.4 | NP_004166.1 | 1 - 2 | 425 | 71 | |
| NR_103819.1 | 1 - 2 | 425 | 71 | |||
| GenBank mRNA | AK296173.1 | - | 1 - 2 | 68 | 71 | |
| BC000457.2 | - | 1 - 2 | 81 | 2822042 | 71 | |
| BC003150.1 | - | 1 - 2 | 81 | 2822042 | 71 | |
| BC034447.1 | - | 1 - 2 | 108 | 71 | ||
| BF344644.1 | - | 1 - 2 | 420 | 71 | ||
| BM928709.1 | - | 1 - 2 | 373 | 71 | ||
| CN286602.1 | - | 1 - 2 | 65 | 71 | ||
| CR456583.1 | - | 1 - 2 | 101 | 71 | ||
| U15009.1 | - | 1 - 2 | 72 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001459.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK296173.1,BC000457.2,BC003150.1,CR456583.1,NM_004175.4,U15009.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal complex assembly
spliceosomal snRNP assembly
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
protein methylation
histone mRNA metabolic process
RNA splicing
nuclear import
protein binding
enzyme binding
U2 snRNA binding
poly(A) RNA binding
telomerase RNA binding
histone pre-mRNA DCP binding
U7 snRNA binding
spliceosomal complex assembly
spliceosomal snRNP assembly
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
protein methylation
histone mRNA metabolic process
RNA splicing
nuclear import
protein binding
enzyme binding
U2 snRNA binding
poly(A) RNA binding
telomerase RNA binding
histone pre-mRNA DCP binding
U7 snRNA binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22002829.hg.1 | TC22001459.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002829.hg.1 | ENST00000215829.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002829.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002835.hg.1 | TC22001459.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002835.hg.1 | ENST00000215829.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002835.hg.1 | ENST00000402849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002835.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002828.hg.1 | TC22001459.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002828.hg.1 | ENST00000215829.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002828.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002834.hg.1 | TC22001459.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002834.hg.1 | ENST00000215829.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002834.hg.1 | ENST00000402849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002834.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | TC2200009235.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168011.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009235.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168008.hg.1 | TC2200009235.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200168008.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168008.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168008.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168008.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168008.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168008.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168008.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168008.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | TC2200009235.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | ENST00000402849.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168014.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168009.hg.1 | TC2200009235.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200168009.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168009.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168009.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168009.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168009.hg.1 | NR_103819.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168009.hg.1 | ENST00000215829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168009.hg.1 | ENST00000468770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168009.hg.1 | NM_001278656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168009.hg.1 | NM_004175.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168009.hg.1 | NR_103819.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 153780 | - | - |

SNRPD3
Genomic Map