Assay Details
Gene Symbol: INPP5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318502.1 | NP_001305431.1 | 8 - 9 | 2066 | 68 | |
NM_019892.5 | NP_063945.2 | 8 - 9 | 2069 | 68 | ||
XM_017014926.1 | XP_016870415.1 | 8 - 9 | 2108 | 68 | ||
GenBank mRNA | AF187891.1 | - | 8 - 9 | 1770 | 68 | |
AL832846.1 | - | 8 - 9 | 2067 | 68 | ||
BC028032.1 | - | 8 - 9 | 2051 | 5242186 | 68 | |
U45974.1 | - | 2 - 3 | 283 | 68 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
