Assay Details
| Gene Symbol: PARP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017554.2 | NP_060024.2 | 7 - 8 | 3372 | 84 | |
| XM_011512928.2 | XP_011511230.1 | 7 - 8 | 3463 | 84 | ||
| XM_011512929.2 | XP_011511231.1 | 7 - 8 | 3471 | 84 | ||
| GenBank mRNA | AB033094.1 | - | 2 - 3 | 1342 | 84 | |
| AK001770.1 | - | 3 - 4 | 400 | 84 | ||
| AK026003.1 | - | 2 - 3 | 1201 | 84 | ||
| AK304269.1 | - | 7 - 8 | 3340 | 84 | ||
| AY134858.1 | - | 2 - 3 | 2545 | 84 | ||
| BX648758.1 | - | 3 - 4 | 2691 | 84 | ||
| DQ063584.1 | - | 4 - 5 | 2836 | 84 | ||
| DQ063585.1 | - | 4 - 5 | 2836 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000634.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03011447.hg.1 | TC03000634.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011447.hg.1 | ENST00000310290.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011447.hg.1 | ENST00000398157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011447.hg.1 | ENST00000398162.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011447.hg.1 | ENST00000474629.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011447.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160954.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160954.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160954.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160954.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160954.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160954.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160954.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160954.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160954.hg.1 | TC0300008561.hg.1 | Clariom D TC |

PARP14
Genomic Map