Assay Details
| Gene Symbol: PARP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017554.2 | NP_060024.2 | 12 - 13 | 4060 | 101 | |
| XM_011512928.2 | XP_011511230.1 | 12 - 13 | 4151 | 101 | ||
| GenBank mRNA | AB033094.1 | - | 7 - 8 | 2030 | 101 | |
| AK001770.1 | - | 8 - 9 | 1088 | 101 | ||
| AK304269.1 | - | 12 - 13 | 4028 | 101 | ||
| AY134858.1 | - | 7 - 8 | 3233 | 101 | ||
| BX648758.1 | - | 8 - 9 | 3378 | 100 | ||
| DQ063584.1 | - | 9 - 10 | 3524 | 101 | ||
| DQ063585.1 | - | 9 - 10 | 3524 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000634.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011454.hg.1 | TC03000634.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011454.hg.1 | ENST00000310290.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011454.hg.1 | ENST00000398157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011454.hg.1 | ENST00000398162.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011454.hg.1 | ENST00000474629.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011454.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000475640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000483611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | ENST00000494811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008561.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160959.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160959.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160959.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160959.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160959.hg.1 | ENST00000460683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160959.hg.1 | ENST00000474629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160959.hg.1 | ENST00000474669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160959.hg.1 | NM_017554.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160959.hg.1 | TC0300008561.hg.1 | Clariom D TC |

PARP14
Genomic Map