Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055727.hg.1 |
TC01003623.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055727.hg.1 |
ENST00000318130.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055727.hg.1 |
ENST00000367512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055727.hg.1 |
NM_025191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226507.hg.1 |
ENST00000318130.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226507.hg.1 |
ENST00000367512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226507.hg.1 |
ENST00000466606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226507.hg.1 |
ENST00000474725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226507.hg.1 |
NM_025191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226507.hg.1 |
ENST00000318130.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226507.hg.1 |
ENST00000367512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226507.hg.1 |
ENST00000466606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226507.hg.1 |
ENST00000474725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226507.hg.1 |
NM_025191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226507.hg.1 |
TC0100016676.hg.1 |
Clariom D TC |