Assay Details
| Gene Symbol: SLC30A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144884.1 | NP_001138356.1 | 1 - 2 | 263 | 66 | |
| NM_133496.4 | NP_598003.2 | 1 - 2 | 261 | 66 | ||
| XM_011540779.2 | XP_011539081.1 | 1 - 2 | 243 | 66 | ||
| XM_017000400.1 | XP_016855889.1 | 1 - 2 | 261 | 66 | ||
| XM_017000401.1 | XP_016855890.1 | 1 - 2 | 261 | 66 | ||
| GenBank mRNA | AF233345.1 | - | 1 - 2 | 91 | 66 | |
| AF529197.1 | - | 1 - 2 | 76 | 66 | ||
| AK313638.1 | - | 1 - 2 | 244 | 66 | ||
| AY094606.1 | - | 1 - 2 | 76 | 66 | ||
| BC064692.1 | - | 1 - 2 | 217 | 30330665 | 66 | |
| BX537375.1 | - | 1 - 2 | 263 | 66 | ||
| CR936806.1 | - | 1 - 2 | 257 | 66 | ||
| DA997188.1 | - | 1 - 2 | 261 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000905.hg
Additional Information:
This assay is in the 5'UTR region of the XM_011540779.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01014264.hg.1 | TC01000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014264.hg.1 | ENST00000370112.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014264.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014264.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370111.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370111.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370111.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014266.hg.1 | TC01000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014266.hg.1 | ENST00000357650.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014266.hg.1 | ENST00000370112.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014266.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014266.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014263.hg.1 | TC01000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014263.hg.1 | ENST00000370112.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014263.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014263.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014265.hg.1 | TC01000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014265.hg.1 | ENST00000357650.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014265.hg.1 | ENST00000370112.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014265.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014265.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165710.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165710.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165710.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165710.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165710.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165710.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165710.hg.1 | TC0100009226.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100165711.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165711.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165711.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165711.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165711.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165711.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165711.hg.1 | TC0100009226.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370111.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370111.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370111.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009226.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165713.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165713.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165713.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165713.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165713.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165713.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165713.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165713.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165713.hg.1 | TC0100009226.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100165714.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165714.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165714.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165714.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165714.hg.1 | ENST00000357650.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165714.hg.1 | ENST00000370112.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165714.hg.1 | NM_001144884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165714.hg.1 | NM_133496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165714.hg.1 | TC0100009226.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 145225 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 172590 | - | - |

SLC30A7
Genomic Map