Assay Details
| Gene Symbol: PET100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033242.1 | 4 - 5 | 281 | 140 | ||
| GenBank mRNA | CB119056.1 | - | 4 - 5 | 272 | 140 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000131.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P391689 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 13493 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 55816 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P391689 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873469 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873471 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485897 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873474 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873470 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873473 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485895 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873468 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873464 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873465 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873459 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873467 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873462 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485889 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485888 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873461 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873463 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873466 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17104476 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873460 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873472 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485894 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873474 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17104476 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873462 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31485888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873472 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873473 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31485895 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873463 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873474 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873470 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873467 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873461 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485894 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873464 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873460 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873473 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485889 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873462 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873471 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873472 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873459 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485897 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873469 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873465 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873468 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485895 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485888 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17104476 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10873466 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31485888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17104476 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873474 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31485895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873472 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873462 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873473 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10873465 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 226006_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000601406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000601406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000601829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000601829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011655.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002048.hg.1 | ENST00000456958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002048.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002048.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002048.hg.1 | TC19000131.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | TC1900011655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199200.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | TC1900011655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199198.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000601406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000601406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000601829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000601829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011655.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | NR_033242.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | TC1900011655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | ENST00000456958.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | ENST00000594797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | ENST00000598540.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | ENST00000600836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | ENST00000623154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | NM_001171155.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199199.hg.1 | NR_033242.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 226006_at | - | - |
| Human Genome U133 Plus 2.0 Array | 226006_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226006_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226006_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226006_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226006_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226006_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226006_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226006_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226006_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226006_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226006_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226006_at2 | - | - |
| Human Genome U95 Set | 55586_at | - | - |
| Human Genome U95 Set | 55586_PM_at | - | - |
| Human Genome U133 Set | 226006_PM_at | - | - |
| Human Genome U133 Set | 226006_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A012137 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A003560 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A012137 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A003560 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 181271 | - | - |

PET100
Genomic Map