Assay Details
| Gene Symbol: PET100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171155.1 | NP_001164626.1 | 2 - 3 | 167 | 94 | |
| NR_033242.1 | 2 - 3 | 167 | 184 | |||
| GenBank mRNA | AW009489.1 | - | 2 - 3 | 94 | ||
| BC150496.1 | - | 2 - 3 | 145 | 94 | ||
| BU740873.1 | - | 2 - 3 | 94 | |||
| CB119056.1 | - | 2 - 3 | 158 | 184 | ||
| CD641681.1 | - | 2 - 3 | 161 | 94 | ||
More Information
Set Membership: |
|

PET100
Genomic Map