| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000413332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000439488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
ENST00000442673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006629.hg.1 |
NR_024455.1 |
Refseq |