Assay Details
| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199051.2 | NP_950252.1 | 1 - 2 | 434 | 134 | |
| XM_017001127.1 | XP_016856616.1 | 1 - 2 | 549 | 134 | ||
| XM_017001129.1 | XP_016856618.1 | 1 - 2 | 434 | 134 | ||
| GenBank mRNA | AB111893.1 | - | 1 - 2 | 181 | 134 | |
| AK123024.1 | - | 1 - 2 | 124 | 134 | ||
| DA809835.1 | - | 1 - 2 | 434 | 134 | ||
| DC330008.1 | - | 1 - 2 | 278 | 134 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003644.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB111893.1,AK123024.1,NM_199051.2,XM_017001127.1,XM_017001129.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000445957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000445957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000445957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000463404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000463404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000633243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | ENST00000633243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055990.hg.1 | TC01003644.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055990.hg.1 | ENST00000367462.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055990.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055997.hg.1 | TC01003644.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055997.hg.1 | ENST00000367462.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055997.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055992.hg.1 | TC01003644.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055992.hg.1 | ENST00000367462.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055992.hg.1 | ENST00000484105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055992.hg.1 | ENST00000534846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055992.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226966.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226966.hg.1 | ENST00000445957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226966.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226966.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226966.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226966.hg.1 | ENST00000445957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226966.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226966.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226966.hg.1 | TC0100016755.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226971.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226971.hg.1 | ENST00000633243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226971.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226971.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226971.hg.1 | ENST00000633243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226971.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226971.hg.1 | TC0100016755.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226964.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226964.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226964.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226964.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226964.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226964.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226964.hg.1 | TC0100016755.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226965.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226965.hg.1 | ENST00000445957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226965.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226965.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226965.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226965.hg.1 | ENST00000445957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226965.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226965.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226965.hg.1 | TC0100016755.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226972.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226972.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226972.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226972.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226972.hg.1 | TC0100016755.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226963.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226963.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226963.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226963.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226963.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226963.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226963.hg.1 | TC0100016755.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000367462.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000445957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000445957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000445957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000463404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000463404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000631494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000633243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | ENST00000633243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016755.hg.1 | NM_199051.2 | Refseq |

BRINP3
Genomic Map