Assay Details
| Gene Symbol: CBX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303494.1 | NP_001290423.1 | 1 - 2 | 216 | 68 | |
| NM_014292.4 | NP_055107.3 | 1 - 2 | 216 | 68 | ||
| GenBank mRNA | AK094829.1 | - | 1 - 2 | 131 | 68 | |
| AK293035.1 | - | 1 - 2 | 98 | 68 | ||
| BC012111.1 | - | 1 - 2 | 95 | 4554483 | 68 | |
| BC064900.1 | - | 1 - 2 | 83 | 6009294 | 68 | |
| CD625491.1 | - | 1 - 2 | 135 | 68 | ||
| HY151719.1 | - | 1 - 2 | 180 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000799.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22013570.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013570.hg.1 | ENST00000407418.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013570.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013570.hg.1 | TC22000799.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013569.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013569.hg.1 | ENST00000407418.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013569.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013569.hg.1 | TC22000799.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013571.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013571.hg.1 | ENST00000407418.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013571.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013571.hg.1 | TC22000799.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013572.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013572.hg.1 | ENST00000407418.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013572.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013572.hg.1 | TC22000799.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013575.hg.1 | ENST00000407418.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013575.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013575.hg.1 | TC22000799.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163287.hg.1 | TC2200008719.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200163287.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163287.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163287.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163287.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163287.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163287.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163287.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163287.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163288.hg.1 | TC2200008719.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200163288.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163288.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163288.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163288.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163288.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163288.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008719.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163282.hg.1 | TC2200008719.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200163282.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163282.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163282.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163282.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163282.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163282.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163282.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163282.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163282.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163282.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163283.hg.1 | TC2200008719.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200163283.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163283.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163283.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163283.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163283.hg.1 | NM_014292.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163283.hg.1 | ENST00000216083.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163283.hg.1 | ENST00000407418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163283.hg.1 | ENST00000469420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200163283.hg.1 | NM_001303494.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200163283.hg.1 | NM_014292.4 | Refseq |

CBX6
Genomic Map